Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LP 12 hydrochloride, Tocris Bioscience™

Catalog No. 292510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-251-0 10 mg
29-255-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-251-0 Supplier R&D Systems Supplier No. 2925/10
This item is not returnable. View return policy

5-HT7 agonist

5-HT7 receptor agonist displaying selectivity over D2, 5-HT1A and 5-HT2A receptors (Ki values are 0.22, 7.3, 52.7 and 326 nM respectively). Induces relaxation of substance P-induced contractions in guinea pig ileum (EC50 = 1.77 μM).

Chemical Identifiers

CAS 1185136-22-4
Molecular Formula C32H40ClN3O
Molecular Weight (g/mol) 518.142
InChI Key XNDUOTNYUWDFKR-UHFFFAOYSA-N
Synonym lp 12 hydrochloride, lp 12 hydrochloride hydrate, 4-2-diphenyl-n-1,2,3,4-tetrahydronaphthalen-1-yl-1-piperazinehexanamide hydrochloride
PubChem CID 56972187
IUPAC Name 6-[4-(2-phenylphenyl)piperazin-1-yl]-N-(1,2,3,4-tetrahydronaphthalen-1-yl)hexanamide;hydrochloride
SMILES C1CC(C2=CC=CC=C2C1)NC(=O)CCCCCN3CCN(CC3)C4=CC=CC=C4C5=CC=CC=C5.Cl

Specifications

Quantity 10 mg
Formula Weight 518.13
Percent Purity >99%
Chemical Name or Material LP 12 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.