Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LP 44, Tocris Bioscience™

Catalog No. 253410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-341-0 10 mg
25-345-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-341-0 Supplier R&D Systems Supplier No. 2534/10
This item is not returnable. View return policy

High affinity 5-HT7 agonist

High affinity 5-HT7 receptor agonist (Ki = 0.22 nM) that displays selectivity over 5-HT1A and 5-HT2A receptors (200- and > 1000-fold respectively). Induces relaxation of substance P-stimulated guinea pig ileum (EC50 = 2.56 μM).

Chemical Identifiers

CAS 824958-12-5
Molecular Formula C27H38ClN3OS
Molecular Weight (g/mol) 488.131
InChI Key DWGKCWWWKHCVDH-UHFFFAOYSA-N
Synonym 4-2-methylthio phenyl-n-1,2,3,4-tetrahydro-1-naphth alenyl-1-piperazinehexanamide hydrochloride, 4-2-methylthio phenyl-n-1,2,3,4-tetrahydro-1-naphthalenyl-1-piperazinehexanamide hydrochloride, lp 44 hydrochloride, lp hplc , solid, 4-2-methylthio phenyl-n-1,2,3,4-tetrahydro-1-naphthalenyl-1-piperazinehexanamidehydrochloride, 6-4-2-methylthio phenyl piperazin-1-yl-n-1,2,3,4-tetrahydronaphthalen-1-yl hexanamide hydrochloride
PubChem CID 11225543
IUPAC Name 6-[4-(2-methylsulfanylphenyl)piperazin-1-yl]-N-(1,2,3,4-tetrahydronaphthalen-1-yl)hexanamide;hydrochloride
SMILES CSC1=CC=CC=C1N2CCN(CC2)CCCCCC(=O)NC3CCCC4=CC=CC=C34.Cl

Specifications

Quantity 10 mg
Formula Weight 488.13
Percent Purity >98%
Chemical Name or Material LP 44
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.