Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LP 99
GREENER_CHOICESDP

Catalog No. 54965 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-965 5 mg
54-962-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 54-965 Supplier Tocris Bioscience™ Supplier No. 5496/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective BRD7 and BRD9 inhibitor (Kd values are 99 and 909 nM, respectively)

LP 99 is a selective BRD7 and BRD9 inhibitor (Kd values are 99 and 909 nM, respectively) that exhibits selectivity for BRD7/9 over a panel of 48 bromodomains (at 10 μM). LP 99 inhibits interactions with histones H3.3 and H4 at low micromolar concentrations, and decreases the level of IL-6 secreted from LPS-stimulated THP-1 cells, in vitro. A negative control for LP 99, ent-LP 99 (Cat.No. 5497), is also available.

Specifications

Chemical Name or Material N-[(2R,3S)-2-(4-Chlorophenyl)-1-(1,2-dihydro-1,4-dimethyl-2-oxo-7-quinolinyl)-6-oxo-3-piperidinyl]-2-methyl-1-propanesulfonamide
CAS 1808951-93-0
Quantity 5 mg
Molecular Formula C26H30ClN3O4S
Formula Weight 516.05
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.