Learn More
Invitrogen™ LPA Monoclonal Antibody (8F6A9,8H5C5)

Description
MA5-15474 targets LPA in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15474 immunogen is purified recombinant fragment of LPA (4330-4521) expressed in E. Coli.
Specifications
Specifications
| Antigen | LPA |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 8F6A9,8H5C5, 8H5C5 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | LPA |
| Gene Accession No. | P08519 |
| Gene Alias | AK38; antiangiogenic AK38 protein; apo(a); APOA; apolipoprotein(a); lipoprotein(a); lipoprotein, Lp(a); LP; lp(a); LPA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.