Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144231
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144231 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144231 Supplier Invitrogen™ Supplier No. PA5144231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LPAL2 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

LPAL2 is a gene that encodes a protein called Lipoprotein, Lp(a)-like 2. Lp(a)-like proteins are structurally similar to lipoprotein(a) (Lp(a)), which is a type of lipoprotein that has been implicated in various cardiovascular diseases. The specific function of LPAL2 is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease. However, studies have suggested that LPAL2 may play a role in lipid metabolism and transport, as well as in the regulation of inflammation and oxidative stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPAL2
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene LPAL2
Gene Accession No. Q16609
Gene Alias APOARGC
Gene Symbols LPAL2
Host Species Rabbit
Immunogen Recombinant Human Putative apolipoprotein(a)-like protein 2 protein (52-124AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80350
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.