Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPAR1/LPA1/EDG-2 Rabbit anti-Human, Mouse, Rat, Clone: 3U1Q0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | LPAR1/LPA1/EDG-2 |
---|---|
Clone | 3U1Q0 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
LPAR1/LPA1/EDG-2 Monoclonal antibody specifically detects LPAR1/LPA1/EDG-2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
LPAR1/LPA1/EDG-2 | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
edg-2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, Gpcr26, GPR26, LPA receptor 1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, VZG1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LPAR1/LPA1/EDG-2 (Q92633). MAAISTSIPVISQPQFTAMNEPQCFYNESIAFFYNRSGKHLATEWNTVSKLVMGLGITVCIFIMLANLLVMVAIYVNRRFHFPIYYLMANLAAADFFAGL | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
3U1Q0 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, GPCR, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1902 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title