Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPAR1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984987
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4987 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4987 Supplier Bioss Supplier No. BS2880R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EDG2 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids There are eight known members of the EDG receptor family and they are implicated in mediating growth-related effects such as induction of cellular proliferation, alterations in differentiation and survival, and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis.

Specifications

Antigen LPAR1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LPAR1
Gene Accession No. Q92633
Gene Alias AI326300; Edg2; edg-2; endothelial differentiation lysophosphatidic acid G-protein-coupled receptor 2; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 2; Gpcr26; Gpcr91; GPR26; G-protein coupled receptor 26; Kdt2; LPA receptor 1; LPA1; LPA-1; LPAR1; Lysophosphatidic acid receptor 1; Lysophosphatidic acid receptor Edg-2; Mrec1.3; rec.1.3; Rec1.3; ventricular zone gene 1; Vzg1; VZG-1
Gene Symbols LPAR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EDG2/LPAR1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1902
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.