Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPAR2/LPA2/EDG-4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | LPAR2/LPA2/EDG-4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
LPAR2/LPA2/EDG-4 Polyclonal specifically detects LPAR2/LPA2/EDG-4 in Human samples. It is validated for Western Blot.Specifications
LPAR2/LPA2/EDG-4 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
EDG-4, EDG4FLJ93869, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor4,LPA receptor EDG4, G protein-coupled receptor, LPA-2, LPA2LPA receptor 2, lysophosphatidic acid receptor 2, lysophosphatidic acid receptor EDG4, Lysophosphatidic acid receptor Edg-4 | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human LPAR2/LPA2/EDG-4 (NP_004711.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, DNA Repair, GPCR, Tumor Suppressors | |
PBS with 50% glycerol, pH7.3. | |
9170 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title