Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPAR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533497
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533497 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533497 Supplier Invitrogen™ Supplier No. PA533497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset, Mouse, Bovine, Dog, Bat, Elephant, Horse, Opossum (100%).

EDG7 is a member of the G protein-coupled receptor family, as well as the EDG family of proteins. This protein functions as a cellular receptor for lysophosphatidic acid and mediates lysophosphatidic acid-evoked calcium mobilization. This receptor couples predominantly to G(q/11) alpha proteins.EDG7 has been reported in human prostate, as well as rodent brain, kidney, lung, placenta, prostate, and testis. ESTs have been isolated from a normal prostate library, as well as cancer libraries of the head/neck, ovary, and thyroid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPAR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Lpar3
Gene Accession No. Q9EQ31, Q9UBY5
Gene Alias calcium-mobilizing lysophosphatidic acid receptor LP-A3; Edg7; Edg-7; endothelial cell differentiation gene 7; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 7; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 7; GPCR; HOFNH30; LPA receptor 3; LPA receptor EDG7; lpA3; LP-A3; LPA-3; LPA3 Receptor; Lpar3; Lysophosphatidic acid receptor 3; lysophosphatidic acid receptor Edg-7; putative G protein-coupled receptor snGPCR32; RP4-678I3; snGPCR32
Gene Symbols Lpar3
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from 2nd cytoplasmic domain of human EDG7.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100052775, 23566, 490187, 532829, 65086
Target Species Human, Mouse, Canine, Bovine, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.