Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPAR4/LPA4 Antibody (561203) [DyLight 755], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10217Z
Description
LPAR4/LPA4 Monoclonal antibody specifically detects LPAR4/LPA4 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
LPAR4/LPA4 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2846 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
561203 | |
Flow Cytometry, CyTOF-ready | |
G protein-coupled receptor 23, GPR23P2Y purinoceptor 9, LPA4LPA-4, lysophosphatidic acid receptor 4, P2RY9G-protein coupled receptor 23, P2Y5-LIKE, P2Y5-like receptor, P2Y9LPA receptor 4, Purinergic receptor 9 | |
Human embryonic kidney cell line HEK293-derived transfected with human LPAR4/LPA4, Accession # Q99677 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction