Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPAR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533895
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533895 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533895 Supplier Invitrogen™ Supplier No. PA533895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Bat, Hamster (100%).

EDG4 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids. There are eight known members of the EDG receptor family and they are implicated in mediating growth-related effects such as induction of cellular proliferation, alterations in differentiation and survival, and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis. EDG receptors are developmentally regulated and differ in tissue distribution. They couple to multiple types of G proteins to signal through ras and MAP kinase, rho, phospholipase C, and several protein tyrosine kinases. EDG4 is expressed in testes, ovarian tumor, and leukocyte containing tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPAR4
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Lpar4
Gene Accession No. Q8BLG2, Q99677
Gene Alias 5730485F04Rik; G protein-coupled receptor 23; Gpr23; G-protein coupled receptor 23; LPA receptor 4; LPA4; LPA-4; LPA4 Receptor; Lpar4; Lysophosphatidic acid receptor 4; P2RY9; P2Y purinoceptor 9; P2Y5-LIKE; P2Y5-like receptor; p2y9; Purinergic receptor 9; RGD1563686
Gene Symbols Lpar4
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from N-terminal extracellular domain of human LPAR4 / GPR23.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100752179, 2846, 302378, 78134
Target Species Human, Mouse, Rat, Monkey, Hamster, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.