Learn More
Invitrogen™ LPAR5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532903
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%); Marmoset (94%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Receptor for lysophosphatidic acid (LPA), a mediator of diverse cellular activities.
Specifications
LPAR5 | |
Polyclonal | |
Unconjugated | |
LPAR5 | |
G protein-coupled receptor 92; GPR92; GPR93; G-protein coupled receptor 92; G-protein coupled receptor 93; KPG_010; LPA receptor 5; LPA5; LPA-5; LPAR5; Lysophosphatidic acid receptor 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57121 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9H1C0 | |
LPAR5 | |
Synthetic 17 amino acid peptide from 3rd extracellular domain of human LPAR5 / GPR92. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.