Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPAR5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533900
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533900 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533900 Supplier Invitrogen™ Supplier No. PA533900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LPAR5 Polyclonal Antibody for IHC (P)

Receptor for lysophosphatidic acid (LPA), a mediator of diverse cellular activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPAR5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene LPAR5
Gene Accession No. Q9H1C0
Gene Alias G protein-coupled receptor 92; GPR92; GPR93; G-protein coupled receptor 92; G-protein coupled receptor 93; KPG_010; LPA receptor 5; LPA5; LPA-5; LPAR5; Lysophosphatidic acid receptor 5
Gene Symbols LPAR5
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human LPAR5 / GPR92.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57121
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.