Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPAR5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100949 Supplier Invitrogen™ Supplier No. PA5100949
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR92/GPR93.

Receptor for lysophosphatidic acid (LPA), a mediator of diverse cellular activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPAR5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LPAR5
Gene Accession No. Q9H1C0
Gene Alias G protein-coupled receptor 92; GPR92; GPR93; G-protein coupled receptor 92; G-protein coupled receptor 93; KPG_010; LPA receptor 5; LPA5; LPA-5; LPAR5; Lysophosphatidic acid receptor 5
Gene Symbols LPAR5
Host Species Rabbit
Immunogen A synthesized peptide derived from human LPAR5(Accession Q9H1C0), corresponding to amino acid residues F247-Y297.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57121
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.