Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPAR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115705 Supplier Invitrogen™ Supplier No. PA5115705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total P2RY5.

The protein encoded by this gene belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. This gene aligns with an internal intron of the retinoblastoma susceptibility gene in the reverse orientation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPAR6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene LPAR6
Gene Accession No. P43657, Q4G072, Q8BMC0
Gene Alias 2610302I02Rik; ARWH1; G-protein coupled purinergic receptor P2Y5; HYPT8; LAH3; LPA receptor 6; LPA-6; LPA6 Receptor; LPAR6; Lysophosphatidic acid receptor 6; oleoyl-L-alpha-lysophosphatidic acid recepto-like protein; oleoyl-L-alpha-lysophosphatidic acid receptor; P2ry5; P2Y purinoceptor 5; P2Y5; purinergic receptor (family A group 5); purinergic receptor 5; purinergic receptor P2Y G protein-coupled protein 5; purinergic receptor P2Y, G-protein coupled, 5; RB intron encoded G-protein coupled receptor
Gene Symbols LPAR6
Host Species Rabbit
Immunogen A synthesized peptide derived from human LPAR6(Accession P43657), corresponding to amino acid residues S103-F153.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10161, 67168, 691774
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.