Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPAR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533901
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533901 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533901 Supplier Invitrogen™ Supplier No. PA533901
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey (94%) Mouse, Rat, Hamster, Rabbit (88%) Bat (81%).

The protein encoded by this gene belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. This gene aligns with an internal intron of the retinoblastoma susceptibility gene in the reverse orientation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPAR6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene LPAR6
Gene Accession No. P43657
Gene Alias 2610302I02Rik; ARWH1; G-protein coupled purinergic receptor P2Y5; HYPT8; LAH3; LPA receptor 6; LPA-6; LPA6 Receptor; LPAR6; Lysophosphatidic acid receptor 6; oleoyl-L-alpha-lysophosphatidic acid recepto-like protein; oleoyl-L-alpha-lysophosphatidic acid receptor; P2ry5; P2Y purinoceptor 5; P2Y5; purinergic receptor (family A group 5); purinergic receptor 5; purinergic receptor P2Y G protein-coupled protein 5; purinergic receptor P2Y, G-protein coupled, 5; RB intron encoded G-protein coupled receptor
Gene Symbols LPAR6
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 2nd extracellular domain of human LPAR6 / P2RY5.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10161
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.