Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPCAT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$405.00 - $670.00
Specifications
Antigen | LPCAT1 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
LPCAT1 Polyclonal specifically detects LPCAT1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LPCAT1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
1-acylglycerophosphocholine O-acyltransferase, Acetyl-CoA:lyso-PAF acetyltransferase, Acetyl-CoA:lyso-platelet-activating factor acetyltransferase, acyl-CoA:lysophosphatidylcholine acyltransferase 1, acyltransferase like 2, Acyltransferase-like 2, AYTL2, EC 2.3.1.-, EC 2.3.1.23,1-alkylglycerophosphocholine O-acetyltransferase, EC 2.3.1.67, FLJ12443, FLJ41609, LPC acyltransferase 1, lpcat, LPCAT-1, Lyso-PAF acetyltransferase, lysoPAFAT, LysoPC acyltransferase 1, lysophosphatidylcholine acyltransferase 1, PFAAP3, Phosphonoformate immuno-associated protein 3, regulated by phosphonoformate | |
LPCAT1 | |
IgG | |
Affinity Purified | |
Specificity of human LPCAT1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
79888 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GVSVTDYTFEDCQLALAEGQLRLPADTCLLEFARLVRGLGLKPEKLEKDLDRYSERARMKGGEKIGIAEFAASLEVPVSDLLEDMFSLFDESGSGEVDLRECVVALSV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title