Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPCAT1 Mouse anti-Human, Mouse, Rat, Clone: 8B6E9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | LPCAT1 |
---|---|
Clone | 8B6E9 |
Concentration | 0.79 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6483
![]() ![]() |
Proteintech Group Inc
660441IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Lysophosphatidylcholine (LPC) acyltransferase (LPCAT; EC 2.3.1.23) catalyzes the conversion of LPC to phosphatidylcholine (PC) in the remodeling pathway of PC biosynthesis (Nakanishi et al., 2006Specifications
LPCAT1 | |
0.79 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Acyltransferase like 2, AYTL2, LPC acyltransferase 1, lpcat, LPCAT 1, LPCAT1, Lyso PAF acetyltransferase, LysoPAFAT, LysoPC acyltransferase 1, PFAAP3 | |
Lpcat1 | |
IgG2b | |
Protein A | |
Antibody |
8B6E9 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q1HAQ0, Q3TFD2, Q8NF37 | |
210992, 361467, 79888 | |
LPCAT1 Fusion Protein Ag9060 | |
Primary | |
-20°C | |
LPCAT1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title