Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPCAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532905
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532905 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532905 Supplier Invitrogen™ Supplier No. PA532905
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LPCAT1 Polyclonal Antibody for IHC (P)

PCAT1 acyltransferase (LPCAT; EC 2.3.1.23) catalyzes the conversion of LPC to hosphatidylcholine (PC) in the remodeling pathway of PC biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPCAT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene LPCAT1
Gene Accession No. Q1HAQ0, Q1LWG4, Q3TFD2, Q8NF37
Gene Alias 1-acylglycerophosphocholine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; 2900035H07Rik; acetyl-CoA:lyso-PAF acetyltransferase; Acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholin; acyl-CoA:lysophosphatidylcholine acyltransferase 1; acyltransferase like 2; acyltransferase-like 2; AGPAT10; AGPAT9; Aytl2; BB137372; BC005662; C87117; LPC acyltransferase 1; LPCAT; LPCAT1; LPCAT-1; lyso-PAF acetyltransferase; lysoPAFAT; lysoPC acyltransferase 1; lysophosphatidylcholine acyltransferase 1; mLPCAT1; PFAAP3; Phosphonoformate immuno-associated protein 3; rd11; regulated by phosphonoformate; RGD1311599
Gene Symbols LPCAT1
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from internal region of human LPCAT1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100217397, 210992, 361467, 420974, 488066, 555969, 79888
Target Species Human, Mouse, Rat, Canine, Monkey, Hamster, Pig, Chicken, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.