Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPCAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596311 Supplier Invitrogen™ Supplier No. PA596311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPADTCLLEF ARLVRGLGLK PEKLEKDLDR YSERARMKGG EKIGIAEFAA SLEVPVSDLL EDMFSLFDES GSGEVDLREC VVALSVVCRP ARTLDTIQLA FKMYGAQEDG SVGEGDLSCI LKTALGVAEL TVTDLFRAID QEEKGKITFA DFHRFAEMYP AFAEEYLYPD QTHFESCAET SPAPIPNGFC ADFSPENSDA GRKPVRKKLD; Positive Samples: BT-474, 22Rv1, Hela, A-549, Mouse lung; Cellular Location: Endoplasmic reticulum membrane, Golgi apparatus membrane, Lipid droplet, Single-pass type II membrane protein.

PCAT1 acyltransferase (LPCAT; EC 2.3.1.23) catalyzes the conversion of LPC to hosphatidylcholine (PC) in the remodeling pathway of PC biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPCAT1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LPCAT1
Gene Accession No. Q1HAQ0, Q3TFD2, Q8NF37
Gene Alias 1-acylglycerophosphocholine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; 2900035H07Rik; acetyl-CoA:lyso-PAF acetyltransferase; Acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholin; acyl-CoA:lysophosphatidylcholine acyltransferase 1; acyltransferase like 2; acyltransferase-like 2; AGPAT10; AGPAT9; Aytl2; BB137372; BC005662; C87117; LPC acyltransferase 1; LPCAT; LPCAT1; LPCAT-1; lyso-PAF acetyltransferase; lysoPAFAT; lysoPC acyltransferase 1; lysophosphatidylcholine acyltransferase 1; mLPCAT1; PFAAP3; Phosphonoformate immuno-associated protein 3; rd11; regulated by phosphonoformate; RGD1311599
Gene Symbols LPCAT1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210992, 361467, 79888
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.