Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LPCAT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552481
Description
Immunogen sequence: FALFDRNHDG SIDFREYVIG LAVLCNPSNT EEIIQVAFKL FDVDEDGYIT EEEFSTILQA SLGVPDLDVS GLFKEIAQGD SISYEEFKSF ALKHPEYAKI FTTYLDLQTC HVFSLPKEVQ TTPSTASNKV SPE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
This gene encodes a member of the lysophospholipid acyltransferase family. The encoded enzyme may function in two ways: to catalyze the biosynthesis of platelet-activating factor (1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine) from 1-O-alkyl-sn-glycero-3-phosphocholine, and to catalyze the synthesis of glycerophospholipid precursors from arachidonyl-CoA and lysophosphatidylcholine. The encoded protein may function in membrane biogenesis and production of platelet-activating factor in inflammatory cells. The enzyme may localize to the endoplasmic reticulum and the Golgi. [provided by RefSeq, Feb 2009].
Specifications
LPCAT2 | |
Polyclonal | |
Unconjugated | |
LPCAT2 | |
1-acylglycerol-3-phosphate O-acyltransferase 11; 1-acylglycerophosphocholine O-acyltransferase; 1-AGP acyltransferase 11; 1-AGPAT 11; 1-alkylglycerophosphocholine O-acetyltransferase; A330042H22; Acetyl-CoA:lyso-PAF acetyltransferase; acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholine acyltransferase 2; acyl-CoA:lysophosphatidylcholine acyltransferase member 1; acyltransferase like 1; acyltransferase like 1A; acyltransferase-like 1; acyltransferase-like 1-A; AGPAT11; AYTL1; Aytl1a; LPAAT-alpha; lpafat1; LPC acyltransferase 2; Lpcat2; LPCAT-2; Lpcat2a; lyso-PAF acetyltransferase; LysoPAFAT; lysoPAFAT/LPCAT2; lysoPC acyltransferase 2; lysophosphatidic acid acyltransferase alpha; Lysophosphatidylcholine acyltransferase 2; lyso-platelet-activating factor (PAF) acetyltransferase; RGD1563994 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100359680, 54947 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P0C1Q3, Q7L5N7 | |
LPCAT2 | |
Recombinant protein corresponding to Human LPCAT2. Recombinant protein control fragment (Product #RP-89707). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction