Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LPCAT3 Monoclonal Antibody (6-2#)
Mouse Monoclonal Antibody
$243.00 - $552.50
Specifications
Antigen | LPCAT3 |
---|---|
Clone | 6-2# |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541634
|
Invitrogen™
MA541634 |
200 μL |
Each for $552.50
|
|
|||||
PIMA543675
|
Invitrogen™
MA543675 |
20 μL |
Each for $243.00
|
|
|||||
Description
LPCAT3 Monoclonal Antibody for Western Blot
LPCAT3 (Lysophosphatidylcholine Acyltransferase 3) is a Protein Coding gene. Diseases associated with LPCAT3 include Retinitis Pigmentosa 40 and Fanconi Anemia, Complementation Group D2. Among its related pathways are triacylglycerol biosynthesis and Glycerophospholipid biosynthesis. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring acyl groups and 1-acylglycerophosphocholine O-acyltransferase activity. An important paralog of this gene is MBOAT1.Specifications
LPCAT3 | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q6P1A2 | |
10162 | |
Recombinant protein Lysophosphatidylcholine Acyltransferase 3. The antigen corresponds to amino acid range 122-233 of the target protein. | |
Antibody |
6-2# | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
LPCAT3 | |
Lysophosphatidylcholine Acyltransferase 3 | |
LPCAT3 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title