Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPCAT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589748 Supplier Invitrogen™ Supplier No. PA589748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGAEPGRSRM ISQEEFARQL QLSDPQTVAG AFGYFQQDTK GLVDFRDVAL ALAALDGGRS LEELTRLAFE LFAEEQAEGP NRLLYKDGFS TILHLLLGSP HPAATALHAE LCQAGSSQGL SLCQFQNFSL HDPLYGKLFS TYLRPPHTSR GTSQTPNASS PGNPTALANG TVQAPKQKGD; Positive Samples: HT-29, HeLa, HepG2, Mouse brain, Mouse testis; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.

Members of the 1-acylglycerol-3-phosphate O-acyltransferase family, such as AGPAT7, catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA), a precursor in the biosynthesis of all glycerolipids. Both LPA and PA are involved in signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPCAT4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LPCAT4
Gene Accession No. Q643R3, Q6NVG1
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 7; 1-acylglycerol-3-phosphate O-acyltransferase 7 (lysophosphatidic acid acyltransferase, eta); 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphoserine O-acyltransferase; 1-AGP acyltransferase 7; 1-AGPAT 7; 1-alkenylglycerophosphoethanolamine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; acyl-CoA:lysophosphatidylethanolamine acyltransferase 2; -acylglycerol-3-phosphate O-acyltransferase 7 (lysophosphatidic acid acyltransferase, eta); acyltransferase like 3; acyltransferase-like 3; AGPAT7; AI505034; Aytl3; LPAAT-eta; LPCAT4; LPEAT2; Lysophosphatidylcholine acyltransferase 4; lysophosphatidylethanolamine acyltransferase 2; Lysophospholipid acyltransferase LPCAT4; plasmalogen synthase; PLSC domain containing protein; RGD1311177
Gene Symbols LPCAT4
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254531, 99010
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.