Learn More
MilliporeSigma™ Supelco™ LpDNPH S10 Cartridge, Bed wt. 350 mg
Shop All MilliporeSigma Supelco Products
Click to view available options
Type:
S10 Cartridge
SPE Cartridge
Quantity:
10/Pk.
50/Pk.
Packaging:
1/Zip Bag
Bulk Packed
Description
The design of the S10 cartridge makes it easy to use in the field and laboratory. Reusable adaptors are available for connecting the cartridge to the sampling pump. The built in reservoir eliminates the need to attach a syringe for sample extraction/elution.
Specifications
Specifications
Basis Weight | 350 mg |
Packaging | Bulk Packed |
Quantity | 50/Pk. |
Type | SPE Cartridge |
Volume (Metric) | 3 mL |