Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LPGAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103561
Description
Antibody detects endogenous levels of total LPGAT1.
LPGAT1 is a lysophoshatidylglycerol (LPG) specific acyltransferase that recognizes various acyl-CoAs and LPGs as substrates but demonstrates a clear preference for long chain saturated fatty acyl-CoAs and oleoyl-CoA as acyl donors. It prefers oleoyl-LPG over palmitoyl-LPG as an acyl receptor and oleoyl-CoA over lauroyl-CoA as an acyl donor.
Specifications
LPGAT1 | |
Polyclonal | |
Unconjugated | |
LPGAT1 | |
Acyl-CoA:lysophosphatidylglycerol acyltransferase 1; AI649174; AW112037; BC013667; FAM34A; FAM34A1; family with sequence similarity 34, member A; KIAA0205; Lpgat1; lysophosphatidylglycerol acyltransferase 1; NET8 | |
Rabbit | |
Affinity chromatography | |
RUO | |
226856, 9926 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q91YX5, Q92604 | |
LPGAT1 | |
A synthesized peptide derived from human LPGAT1(Accession Q92604), corresponding to amino acid residues H144-T194. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction