Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LPGAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113143
Description
LPGAT1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
LPGAT1 is a lysophoshatidylglycerol (LPG) specific acyltransferase that recognizes various acyl-CoAs and LPGs as substrates but demonstrates a clear preference for long chain saturated fatty acyl-CoAs and oleoyl-CoA as acyl donors. It prefers oleoyl-LPG over palmitoyl-LPG as an acyl receptor and oleoyl-CoA over lauroyl-CoA as an acyl donor.
Specifications
LPGAT1 | |
Polyclonal | |
Unconjugated | |
LPGAT1 | |
Acyl-CoA:lysophosphatidylglycerol acyltransferase 1; AI649174; AW112037; BC013667; FAM34A; FAM34A1; family with sequence similarity 34, member A; KIAA0205; Lpgat1; lysophosphatidylglycerol acyltransferase 1; NET8 | |
Rabbit | |
Protein G | |
RUO | |
9926 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4.14 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q92604 | |
LPGAT1 | |
Recombinant Human Acyl-CoA:lysophosphatidylglycerol acyltransferase 1 protein (151-341AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction