Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPHN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533903
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533903 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533903 Supplier Invitrogen™ Supplier No. PA533903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LPHN1 Polyclonal Antibody for IHC (P)

Adgrl1 (Latrophilin-1) is a calcium-independent receptor of high affinity for alpha-latrotoxin, an excitatory neurotoxin present in black widow spider venom which triggers massive exocytosis from neurons and neuroendocrine cells. Adgrl1 is a receptor for TENM2 that mediates heterophilic synaptic cell-cell contact and postsynaptic specialization, and may be implicated in the regulation of exocytosis. The Adgrl1 gene encodes a member of the latrophilin subfamily of G-protein coupled receptors (GPCR). Latrophilins may function in both cell adhesion and signal transduction. In experiments with non-human species, endogenous proteolytic cleavage within a cysteine-rich GPS (G-protein-coupled-receptor proteolysis site) domain resulted in two subunits (a large extracellular N-terminal cell adhesion subunit and a subunit with substantial similarity to the secretin/calcitonin family of GPCRs) being non-covalently bound at the cell membrane. Adgrl1 has been shown to recruit the neurotoxin from black widow spider venom, alpha-latrotoxin, to the synapse plasma membrane. Alternative splicing results in multiple variants encoding distinct isoforms of Adgrl1. Diseases associated with ADGRL1 include Dermatophytosis and Retinal Degeneration, Late-Onset, Autosomal Dominant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPHN1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Adgrl1
Gene Accession No. O88917, O94910, O97831, Q80TR1
Gene Alias 2900070I05Rik; ADGRL1; Adhesion G protein-coupled receptor L1; AI182398; calcium-independent alpha-latrotoxin receptor 1; Cirl; Cirl1; CIRL-1; CL1; CL1BA; CLIBA; KIAA0821; latrophilin 1; latrophilin-1; LEC2; Lectomedin-2; Lphh2; LPHN1; mKIAA0821
Gene Symbols Adgrl1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-terminal extracellular domain of human LPHN1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100359196, 100514215, 100760780, 22859, 330814, 610349, 65096, 788252
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.