Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPHN1 Polyclonal Antibody
LPHN1 Polyclonal Antibody
Supplier: Thermo Scientific PA575811
Description
LPHN1 Polyclonal Antibody for Western Blot

Specifications
LPHN1 | |
Polyclonal | |
Unconjugated | |
Adgrl1 | |
2900070I05Rik; ADGRL1; Adhesion G protein-coupled receptor L1; AI182398; calcium-independent alpha-latrotoxin receptor 1; Cirl; Cirl1; CIRL-1; CL1; CL1BA; CLIBA; KIAA0821; latrophilin 1; latrophilin-1; LEC2; Lectomedin-2; Lphh2; LPHN1; mKIAA0821 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22859 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 15 mM sodium azide; pH 7.2 | |
O94910 | |
Adgrl1 | |
Synthetic peptide corresponding to residues in Human LPHN1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction