Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPHN2 Polyclonal Antibody
LPHN2 Polyclonal Antibody
Supplier: Thermo Scientific PA575812
Description
LPHN2 Polyclonal Antibody for Western Blot

Specifications
LPHN2 | |
Polyclonal | |
Unconjugated | |
ADGRL2 | |
Adgrl2; Adhesion G protein-coupled receptor L2; AI450192; Calcium-independent alpha-latrotoxin receptor 2; calcium-independent alpha-latrotoxin receptor homolog 2; Cirl2; Cirl-2; Cl2; Cl2aa; Cl2ac; Gm619; KIAA0786; latrophilin 1; latrophilin 2; latrophilin homolog 1; Latrophilin-2; LEC1; Lectomedin-1; LPHH1; Lphn2; mKIAA0786 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23266 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O95490 | |
ADGRL2 | |
Synthetic peptide corresponding to residues in Human LPHN2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction