Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPHN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA565359
Description
Immunogen sequence: PVKPVIGGSSS EDDAIVADAS SLMHSDNPGL ELHHKELEAP LIPQRTHSLL YQPQKKVKSE GTDSYVSQLT AEAEDHLQS Highest antigen sequence identity to the following orthologs - mouse 93%, rat 91%.
Calcium-independent receptor of low affinity for alpha-latrotoxin, an excitatory neurotoxin present in black widow spider venom which triggers massive exocytosis from neurons and neuroendocrine cells. Receptor propably implicated in the regulation of exocytosis.
Specifications
LPHN2 | |
Polyclonal | |
Unconjugated | |
ADGRL2 | |
LPHN2; Cl2aa; CIRL2; CL2; Latrophilin 1; Latrophilin-2; Lectomedin-1; Latrophilin 2; Latrophilin homolog 2 (cow); KIAA0786; Beta-latrotoxin receptor; CIRL-2; Latrophilin homolog 1; Latrotoxin receptor 2; Latrotoxin receptor homolog 2; LEC1; LPHH1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23266 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95490 | |
ADGRL2 | |
Recombit Human LPHN2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction