Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPHN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533906
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533906 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533906 Supplier Invitrogen™ Supplier No. PA533906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LPHN2 Polyclonal Antibody for IHC (P)

The Latrophilin family of G protein-coupled receptors consists of three members, latrophilin-1, 2 and 3, each of which displays distinct tissue distribution and function. Latrophilin-1, the most characterized member of this family, acts as a receptor for a-latrotoxin, a component of venom from the black widow spider. Binding of a-latrotoxin to latrophilin-1 triggers synaptic vesicle exocytosis via both Ca2+-dependent and -independent mechanisms, resulting in vesicle depletion. Latrophilin-1 is abundant in brain and present in endocrine cells. Latrophilin-3 is also brain-specific, whereas latrophilin-2 expression is ubiquitous.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPHN2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRL2
Gene Accession No. O88923, O95490, O97817, Q8JZZ7
Gene Alias Adgrl2; Adhesion G protein-coupled receptor L2; AI450192; Calcium-independent alpha-latrotoxin receptor 2; calcium-independent alpha-latrotoxin receptor homolog 2; Cirl2; Cirl-2; Cl2; Cl2aa; Cl2ac; Gm619; KIAA0786; latrophilin 1; latrophilin 2; latrophilin homolog 1; Latrophilin-2; LEC1; Lectomedin-1; LPHH1; Lphn2; mKIAA0786
Gene Symbols ADGRL2
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-terminal extracellular domain of human LPHN2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100052932, 100343149, 100524685, 171447, 23266, 281278, 490193, 99633
Target Species Human, Mouse, Rat, Canine, Rabbit, Bovine, Pig, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.