Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPHN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533909
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533909 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533909 Supplier Invitrogen™ Supplier No. PA533909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster, Panda, Horse, Opossum, Turkey, Chicken, Platypus (100%).

Plays a role in cell-cell adhesion and neuron guidance via its interactions with FLRT2 and FLRT3 that are expressed at the surface of adjacent cells. Plays a role in the development of glutamatergic synapses in the cortex. Important in determining the connectivity rates between the principal neurons in the cortex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPHN3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRL3
Gene Accession No. Q80TS3, Q9HAR2, Q9Z173
Gene Alias 5430402I23Rik; ADGRL3; Adhesion G protein-coupled receptor L3; calcium-independent alpha-latrotoxin receptor 3; calcium-independent alpha-latrotoxin receptor homolog 3; Cirl3; CIRL-3; Cl3; D130075K09Rik; Gm1379; Kiaa0768; latrophilin 3; latrophilin homolog 3 (cow); latrophilin-3; LEC3; lectomedin 3; lectomedin-3; Lph3; LPHN3; mKIAA0768
Gene Symbols ADGRL3
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human LPHN3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100056929, 100757140, 170641, 23284, 319387, 422616
Target Species Human, Mouse, Rat, Monkey, Hamster, Horse, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.