Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPHN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567880
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567880 Supplier Invitrogen™ Supplier No. PA567880
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Plays a role in cell-cell adhesion and neuron guidance via its interactions with FLRT2 and FLRT3 that are expressed at the surface of adjacent cells. Plays a role in the development of glutamatergic synapses in the cortex. Important in determining the connectivity rates between the principal neurons in the cortex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPHN3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADGRL3
Gene Accession No. Q80TS3, Q9HAR2, Q9Z173
Gene Alias 5430402I23Rik; ADGRL3; Adhesion G protein-coupled receptor L3; calcium-independent alpha-latrotoxin receptor 3; calcium-independent alpha-latrotoxin receptor homolog 3; Cirl3; CIRL-3; Cl3; D130075K09Rik; Gm1379; Kiaa0768; latrophilin 3; latrophilin homolog 3 (cow); latrophilin-3; LEC3; lectomedin 3; lectomedin-3; Lph3; LPHN3; mKIAA0768
Gene Symbols ADGRL3
Host Species Rabbit
Immunogen Synthesized peptide derived from human LPHN3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170641, 23284, 319387
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.