Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPHN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $371.50
Specifications
Antigen | LPHN3 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145296
|
Invitrogen™
PA5145296 |
20 μL |
Each for $191.80
|
|
|||||
FEPA5145295
|
Invitrogen™
PA5145295 |
200 μL |
Each for $371.50
|
|
|||||
Description
Plays a role in cell-cell adhesion and neuron guidance via its interactions with FLRT2 and FLRT3 that are expressed at the surface of adjacent cells. Plays a role in the development of glutamatergic synapses in the cortex. Important in determining the connectivity rates between the principal neurons in the cortex.Specifications
LPHN3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q80TS3, Q9HAR2, Q9Z173 | |
170641, 23284, 319387 | |
Recombinant Human LPHN3 protein, Gln1105-Leu1447 (Accession #Q9HAR2). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
5430402I23Rik; ADGRL3; Adhesion G protein-coupled receptor L3; calcium-independent alpha-latrotoxin receptor 3; calcium-independent alpha-latrotoxin receptor homolog 3; Cirl3; CIRL-3; Cl3; D130075K09Rik; Gm1379; Kiaa0768; latrophilin 3; latrophilin homolog 3 (cow); latrophilin-3; LEC3; lectomedin 3; lectomedin-3; Lph3; LPHN3; mKIAA0768 | |
ADGRL3 | |
IgG | |
Protein A | |
ADGRL3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title