Learn More
LPHN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | LPHN3 |
|---|---|
| Concentration | 0.32 mg/mL |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2671
![]()
|
Proteintech Group Inc
200451AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody recognizes all the isoforms of LPHN3 and has no cross reaction with LPHN1 and LPHN2.
Plays a role in cell-cell adhesion and neuron guidance via its interactions with FLRT2 and FLRT3 that are expressed at the surface of adjacent cells. Plays a role in the development of glutamatergic synapses in the cortex. Important in determining the connectivity rates between the principal neurons in the cortex.Specifications
| LPHN3 | |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q80TS3, Q9HAR2, Q9Z173 | |
| 170641, 23284, 319387 | |
| Peptide | |
| Primary | |
| -20°C | |
| ADGRL3 |
| 0.32 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CIRL 3, CIRL3, KIAA0768, latrophilin 3, LEC3, Lectomedin 3, LPHN3 | |
| Adgrl3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.