Learn More
LPIN1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LPIN1 |
---|---|
Concentration | 267 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8549
![]() ![]() |
Proteintech Group Inc
270261AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The LPIN1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human LPIN1. This antibody recognizes human antigen. The LPIN1 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
LPIN1 | |
Western Blot, Immunoprecipitation, Immunohistochemistry | |
Unconjugated | |
Human | |
IgG |
267 μg/mL | |
Polyclonal | |
Rabbit | |
23175 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.