Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPIN3 Monoclonal Antibody (OTI1D7)
GREENER_CHOICE

Catalog No. PIMA526707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526707 Supplier Invitrogen™ Supplier No. MA526707
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LPIN3 Monoclonal Antibody for Western Blot, IHC (P)

Humans lipodystrophy is characterized by loss of body fat, fatty liver, hypertriglyceridemia, and insulin resistance. Mice carrying mutations in the fatty liver dystrophy gene have similar phenotypes. Through positional cloning, the mouse gene responsible for fatty liver dystrophy was isolated and designated Lpin1. The nuclear protein encoded by Lpin1 was named lipin. Lpin1 mRNA was expressed at high levels in adipose tissue and was induced during differentiation of preadipocytes. These results indicated that lipin is required for normal adipose tissue development and provided a candidate gene for human lipodystrophy. Through database searches, mouse and human EST and genomic sequences with similarities to Lpin1 were identified. These included two related mouse genes and three human homologs. Human LPIN1 gene has been mapped to 2p25.; linkages of fat mass and serum leptin levels to this same region have been noted. Human LPIN2 and LPIN3 mapped to chromosomes 18p11 and 20q11-q12, respectively. The mouse genes encoding Lpin1, Lpin2, and Lpin3 mapped to chromosome 12, 17, and 2, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPIN3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LPIN3
Gene Accession No. Q9BQK8
Gene Alias 9130206L11Rik; AA438110; AV236139; dJ450M14.2; dJ450M14.3; dJ620E11.2; lipin 3; lipin 3-like; lipin-3; Lipin-3-like; LIPN3L; Lpin3; mKIAA4023; Phosphatidate phosphatase LPIN3; RP4-620E11.3; SMP2
Gene Symbols LPIN3
Host Species Mouse
Immunogen Full length human recombinant protein of LPIN3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64900
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.