Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LPL Monoclonal Antibody (5D2), HRP
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA546592
This item is not returnable.
View return policy
Description
Predicted molecular weight: 53 kDa.
LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism.
Specifications
LPL | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P06858, P11151, P11152, P11153, P11602, Q06000 | |
LPL | |
Purified bovine milk lipoprotein lipase protein. | |
50 μg | |
Primary | |
Human, Mouse, Rat, Bovine, Chicken, Guinea Pig | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
5D2 | |
HRP | |
LPL | |
adipose lipoprotein lipase; alpha lipase; HDLCQ11; I79_006077; LIPD; lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); lipoprotein lipase precursor; lipoprotein lipase preprotein; Lipoprotein lipase-like protein; LPL; O 1-4-5; phospholipase A1; triacylglycerol lipase | |
Mouse | |
Protein G | |
RUO | |
100135570, 16956, 24539, 280843, 396219, 4023 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction