Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPL Monoclonal Antibody (5D2)
GREENER_CHOICE

Catalog No. PIMA518055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518055 Supplier Invitrogen™ Supplier No. MA518055
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not recognize hepatic lipase and does not react with mouse samples. This target has a predicted molecular weight of 53 kDa.

LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPL
Applications ELISA, Inhibition Assays, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene LPL
Gene Accession No. P06858, P11151, P11153, P11602, Q06000
Gene Alias adipose lipoprotein lipase; alpha lipase; HDLCQ11; I79_006077; LIPD; lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); lipoprotein lipase precursor; lipoprotein lipase preprotein; Lipoprotein lipase-like protein; LPL; O 1-4-5; phospholipase A1; triacylglycerol lipase
Gene Symbols LPL
Host Species Mouse
Immunogen Purified bovine milk lipoprotein lipase protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100135570, 24539, 280843, 396219, 4023
Target Species Human, Rat, Bovine, Chicken, Guinea Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.