Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPL Monoclonal Antibody (OTI4G7), TrueMAB™, OriGene
SDP

Catalog No. 502540660
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0660 30 μL
50-167-4663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0660 Supplier OriGene Supplier No. TA503880S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LPL Monoclonal antibody specifically detects LPL in Canine, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen LPL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4G7
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Usp10
Gene Accession No. P52479, Q14694, Q3KR59
Gene Alias adipose lipoprotein lipase; alpha lipase; HDLCQ11; I79_006077; LIPD; lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); lipoprotein lipase precursor; lipoprotein lipase preprotein; Lipoprotein lipase-like protein; LPL; O 1-4-5; phospholipase A1; triacylglycerol lipase
Host Species Mouse
Immunogen Full length human recombinant protein of human USP10 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22224, 307905, 479625, 9100
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.