Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPL Monoclonal Antibody (OTI4H3), TrueMAB™, OriGene
SDP

Catalog No. 502540709
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0709 30 μL
50-167-4592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0709 Supplier OriGene Supplier No. TA503787S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LPL Monoclonal antibody specifically detects LPL in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen LPL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LPL
Gene Accession No. P06858
Gene Alias adipose lipoprotein lipase; alpha lipase; HDLCQ11; I79_006077; LIPD; lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); lipoprotein lipase precursor; lipoprotein lipase preprotein; Lipoprotein lipase-like protein; LPL; O 1-4-5; phospholipase A1; triacylglycerol lipase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 28-475 of human LPL produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4023
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.