Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPL Monoclonal Antibody (OTI6B4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503784S


Description
LPL Monoclonal antibody specifically detects LPL in Human samples. It is validated for Western BlotSpecifications
LPL | |
Monoclonal | |
0.61 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P06858 | |
Mouse | |
Affinity chromatography | |
RUO | |
4023 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI6B4 | |
Unconjugated | |
LPL | |
adipose lipoprotein lipase; alpha lipase; HDLCQ11; I79_006077; LIPD; lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); lipoprotein lipase precursor; lipoprotein lipase preprotein; Lipoprotein lipase-like protein; LPL; O 1-4-5; phospholipase A1; triacylglycerol lipase | |
Human recombinant protein fragment corresponding to amino acids 28-475 of human LPL produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction