Learn More
LPL Mouse anti-Human, Clone: OTI6A10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503791

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism.Specifications
LPL | |
Monoclonal | |
Unconjugated | |
LPL | |
HDLCQ11, LIPD | |
Mouse | |
Affinity Chromatography | |
RUO | |
4023 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI6A10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P06858 | |
LPL | |
Human recombit protein fragment corresponding to amino acids 28-475 of human LPL produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.