Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LPL Recombinant Rabbit Monoclonal Antibody (10E10P9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535444
Description
Immunogen sequence: IPFTLPEVST NKTYSFLIYT EVDIGELLML KLKWKSDSYF SWSDWWSSPG FAIQKIRVKA GETQKKVIFC SREKVSHLQK GKAPAVFVKC HDKSLNKKSG.
LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism.
Specifications
LPL | |
Recombinant Monoclonal | |
0.6 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06858, P11152 | |
LPL | |
A synthetic peptide corresponding to a sequence within amino acids 376-475 of human Lipoprotein lipase (LPL) (P06858). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
10E10P9 | |
Unconjugated | |
LPL | |
adipose lipoprotein lipase; alpha lipase; HDLCQ11; I79_006077; LIPD; lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); lipoprotein lipase precursor; lipoprotein lipase preprotein; Lipoprotein lipase-like protein; LPL; O 1-4-5; phospholipase A1; triacylglycerol lipase | |
Rabbit | |
Affinity chromatography | |
RUO | |
16956, 4023 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction