Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPLUNC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980609
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0609 Supplier Bioss Supplier No. BS0976R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene may be involved in the innate immune response to bacterial exposure in the mouth, nasal cavities, and lungs. The encoded protein is secreted and is a member of the BPI/LBP/PLUNC protein superfamily. This gene is found with other members of the superfamily in a cluster on chromosome 20.

Specifications

Antigen LPLUNC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BPIFB1
Gene Accession No. Q8TDL5
Gene Alias BPI fold containing family B member 1; BPI fold containing family B, member 1; BPI fold-containing family B member 1; BPIFB1; C20orf114; long palate, lung and nasal epithelium carcinoma associated 1; Long palate, lung and nasal epithelium carcinoma-associated protein 1; Lplunc1; UNQ706/PRO1357; VEMSGP; von Ebner minor salivary gland protein; von Ebner minor salivary protein
Gene Symbols BPIFB1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LPLUNC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92747
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.