Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA146352
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA146352 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA146352 Supplier Invitrogen™ Supplier No. PA146352
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The target sequence has 81% sequence homology with sheep and 73% sequence homology with mouse and rat.

This gene encodes an oxidoreductase secreted from salivary, mammary, and other mucosal glands that functions as a natural antibacterial agent. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPO
Applications Western Blot
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.1% sodium azide
Gene LPO
Gene Accession No. P22079, P80025
Gene Alias 5830499B15Rik; airway lactoperoxidase; EC 1.11.17 antibody; lactoperoxidase; lactoperoxydase; Lpo; LPO antibody; MGC129990; MGC129991; Salivary peroxidase; SAPX; SAPX antibody; SPO; SPO antibody
Gene Symbols LPO
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human Lactoperoxidase protein (within residues 300-400).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280844, 4025
Target Species Human, Monkey, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.