Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109923 Supplier Thermo Scientific Supplier No. PA5109923
Add to Cart
Add to Cart

LPO Polyclonal Antibody

LPO Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen LPO
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LPO
Gene Accession No. P22079
Gene Alias 5830499B15Rik; airway lactoperoxidase; EC 1.11.17 antibody; lactoperoxidase; lactoperoxydase; Lpo; LPO antibody; MGC129990; MGC129991; Salivary peroxidase; SAPX; SAPX antibody; SPO; SPO antibody
Gene Symbols LPO
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 27-280 of human LPO (NP_006142.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4025
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.