Learn More
Invitrogen™ LPO Polyclonal Antibody, HRP

Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | LPO |
| Applications | Dot Blot, ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 10 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 1% BSA and 0.01% gentamicin sulfate; pH 7.2 |
| Gene | LPO |
| Gene Accession No. | P80025 |
| Gene Alias | 5830499B15Rik; airway lactoperoxidase; EC 1.11.17 antibody; lactoperoxidase; lactoperoxydase; Lpo; LPO antibody; MGC129990; MGC129991; Salivary peroxidase; SAPX; SAPX antibody; SPO; SPO antibody |
| Gene Symbols | LPO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.