Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPPR1/PRG3 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12150R

Description
This gene encodes a member of the plasticity-related gene (PRG) family. Members of the PRG family mediate lipid phosphate phosphatase activity in neurons and are known to be involved in neuronal plasticity. The protein encoded by this gene does not perform its function through enzymatic phospholipid degradation. This gene is strongly expressed in brain. It shows dynamic expression regulation during brain development and neuronal excitation. Alternatively spliced transcript variants encoding the same protein have been observed. Possesses similar cytotoxic and cytostimulatory activities to PRG2/MBP. In vitro, stimulates neutrophil superoxide production and IL8 release, and histamine and leukotriene C4 release from basophils.Specifications
LPPR1/PRG3 | |
Polyclonal | |
Unconjugated | |
PRG3 | |
Eosinophil major basic protein 2; Eosinophil major basic protein homolog; major basic protein 2; Mbp2; MBPH; Prepro-major basic protein homolog; Prepro-MBPH; Prg3; Proteoglycan 3; proteoglycan 3, pro eosinophil major basic protein 2; UNQ486/PRO1002 | |
Rabbit | |
Protein A | |
RUO | |
10394, 295706, 53856 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9JL95, Q9Y2Y8 | |
PRG3 | |
KLH conjugated synthetic peptide derived from human LPPR1/PRG3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction