Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPPR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543416 Supplier Invitrogen™ Supplier No. PA543416
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NYTALGCLPP SPDRPGPDRF VTDQGACAGS PSLVAAARRA FPCKDAALCA Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

LPPR2 is a multi-pass membrane protein and belongs to the PA-phosphatase related phosphoesterase family. LPPR2 shows dynamic expression regulation during brain development and neuronal excitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPPR2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PLPPR2
Gene Accession No. Q96GM1
Gene Alias Lipid phosphate phosphatase-related protein type 2; Lppr2; MGC137198 protein; phospholipid phosphatase related 2; phospholipid phosphatase-related protein type 2; plasticity related gene 4; plasticity related protein 4; plasticity-related gene 4 protein; PLPPR2; PRG4; PRG-4
Gene Symbols PLPPR2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LPPR2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64748
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.